missing translation for 'onlineSavingsMsg'
Learn More

TIGIT Antibody (TIGIT/3017), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79927G

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215187

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

TIGIT
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
TIGIT
Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary) (Uniprot: Q4951)
0.1 mL
Primary
Store at 4°C in the dark.
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TIGIT/3017
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM
Mouse
Protein A or G purified
RUO
Human
IgG2c κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales