Learn More
Tiaprofenic Acid, TRC
Product Code. 30449327
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30449327 25 mg 25mg
1 options
This item is not returnable. View return policy
Product Code. 30449327 Supplier Toronto Research Chemicals Supplier No. TRCT43700025MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tiaprofenic acid
CAS 33005-95-7
Molecular Formula C14 H12 O3 S
Quantity 25 mg
Synonym Tiaprofenic acid, (2RS)-2-(5-Benzoylthiophen-2-yl)propanoic acid
InChI Formula InChI=1S/C14H12O3S/c1-9(14(16)17)11-7-8-12(18-11)13(15)10-5-3-2-4-6-10/h2-9H,1H3,(H,16,17)
SMILES CC(C(=O)O)c1ccc(s1)C(=O)c2ccccc2
IUPAC Name 2-(5-benzoylthiophen-2-yl)propanoic acid
Molecular Weight (g/mol) 260.31
Formula Weight 260.0507