Learn More
Tiaprofenic Acid, Mikromol™
Product Code. 30049886
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30049886 100 mg 100mg
1 options
This item is not returnable. View return policy
Product Code. 30049886 Supplier Mikromol Supplier No. MM3322.00
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tiaprofenic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tiaprofenic acid
CAS 33005-95-7
Analyte or Component Names Tiaprofenic acid
Shipping Condition Room Temperature
Molecular Formula C14 H12 O3 S
Certifications/Compliance ISO 17034
Synonym Tiaprofenic acid, (2RS)-2-(5-Benzoylthiophen-2-yl)propanoic acid
InChI Formula InChI=1S/C14H12O3S/c1-9(14(16)17)11-7-8-12(18-11)13(15)10-5-3-2-4-6-10/h2-9H,1H3,(H,16,17)
SMILES CC(C(=O)O)c1ccc(s1)C(=O)c2ccccc2
IUPAC Name 2-(5-benzoylthiophen-2-yl)propanoic acid
Molecular Weight (g/mol) 260.31
Quantity 100 mg
Formula Weight 260.0507 g/mol