missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18304-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.Especificaciones
Thrombospondin 1 | |
Polyclonal | |
Unconjugated | |
THBS1 | |
THBS, THBS 1, THBS1, thrombospondin 1, TSP, TSP 1, TSP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21825, 445442, 7057 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07996, P35441 | |
Thbs1 | |
Peptide | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |