missing translation for 'onlineSavingsMsg'
Learn More

Thrombomodulin / CD141 Mouse, Clone: THBD/1782, enQuireBio™

Mouse Monoclonal Antibody

Marca:  EnquireBio 7056-MSM2-P0

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15930968

  • 199.00€ / 20 microgramos


Fecha estimada de envío
18-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

Thrombomodulin / CD141
Cell Surface and Cytoplasmic
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
THBD
Recombinant human Thrombomodulin protein fragment (aa69-194)
100kDa
Protein A/G Purified
RUO
Primary
7056
Immunohistochemistry
THBD/1782
Unconjugated
NP_000352
Mouse
IgG2a kappa
THP1 Cells. Placenta, Angiosarcoma, Bladder or Cervical Carcinoma
20 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
Documentos