Learn More
S.C.A.T. Europe™ Thread S60/61 SafetyWasteCap
For safe, clean liquid disposal. Ideal for HPLC waste disposal or any liquid waste disposal in the Lab. Easy to use and comes with self-sealing fittings, connectors and funnel, free turning PTFE body and port for the exhaust filter. Offers reassuring protection and efficient work in the laboratory
Marca:  S.C.A.T. Europe™ 108134
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 10222090
  • 489.01€ / cada uno
Fecha estimada de envío
06-11-2019
Añadir a la cesta
Descripción
Descripción
  • Free turning PTFE body
  • Connector for Exhaust Filter
  • Lab safety, optimal protection when collecting liquid waste
  • Resistant against aggressive media
  • Filter system against harmful vapors
  • Flexible system
  • Huge range of accessories
  • Possibility of funnel use
  • Safe disposal of HPLC waste
  • Work performance
  • Personal protection
  • Occupational safety
  • Ergonomics
  • Cost efficiency
  • Technical protective measures
  • Preventing overflowing containers with level control systems
  • Optimal protection when collecting liquid waste
  • Sealed bottles and containers even when tumbling over
  • No need to use local extractors
  • Time saving in changing the waste containers while tubes are fixed
  • Create a comfortable working climate
  • Prevent inhalation of harmful solvent vapors
  • Avoid skin contact with organic solvents
Especificaciones
Especificaciones
Includes: 2 x 3.2 mm OD Capillary Connector. Mechanical Level Control. Safety Funnel with Automatic Closure.
Blue
Polypropylene
Security Cap Thread S60/61