Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Research tool for transport, co-culture, migration and invasion studies. Greriner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion through physical surface (TC) treatment.
Marca:  Greiner Bio-One 662610
Detalles adicionales : Peso : 0.80000kg
 Ver más versiones de este producto
Código de producto. 10443845
  • 595.00€ / paquete de 48
Fecha de envío estimada
del proveedor 04-03-2019
Añadir a la cesta
Descripción
Descripción
  • Clear polystyrene housing with sealed PET capillary pore membrane
  • Hanging geometry provides protection to lower compartment cell layer
  • Simplified pipetting due to self-lift geometry
  • Eccentric in-well positioning allows unequalled access to lower compartment
  • Available in 6, 12 and 24 well format
Especificación
Especificación
Sterile
33.6mm2
0.1 - 0.35mL
1.0μm
For 24 Well
10.4mm
48Pack
24
Polystyrene
2 x 106
Thincert Cell Culture Insert
8.4mm
48Pack
48Pack
Videos