Learn More
Brannan™ Classroom Wall Thermometer Kit
Product Code. 13028573
Change view
Click to view available options
Height (Metric):
380 mm
Unit Size:
1 set
Product Code. Height (Metric) Unit Size
13028573 380 mm 1 set
1 options
This item is not returnable. View return policy
Product Code. 13028573 Supplier Brannan™ Supplier No. 25/702/0
  • ONLINE EXCLUSIVE PRICE
    16.55€
    15.50€ / 1 sets
    Ahorro 1.05€
    6% Descuento
Fecha estimada de envoi: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Brannan™ Classroom Wall Thermometer Kit includes a large, colorful, plastic wall thermometer and chart for recording temperatures.
  • Colorful, easy to read thermometer
  • Laminated recording chart
  • Ideal for educational use
Specifications
Includes Laminated master thermometer chart
Height (Metric) 380 mm
Product Type Wall Thermometer Kit