Learn More
Invitrogen™ Human IL-1RA Recombinant Protein
Recombinant Protein
Marca: Invitrogen™ RP75579
Detalles adicionales : CAS : 69-74-9 Peso : 0.01000kg
Descripción
IL-1 Receptor Antagonist (human) Recombinant Protein, RP-75579, from Invitrogen.
This product contains purified recombinant human IL-1 Receptor Antagonist protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays. This protein has a molecular weight of ∽21.63 kDa Purity >95% by SDS-PAGE.Especificaciones
P18510 | |
20mM tris HCl with 50% glycerol and no preservative; pH 8.0 | |
Human IL1RA | |
100μg | |
N/A | |
RUO | |
IL1RN | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control, Western Blot | |
3557 | |
8.0 | |
E. coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human IL1RA | |
Human | |
Hu | |
Recombinant | |
Recombinant |