Learn More
Thermo Scientific™ Orion™ Ammonium Ion Sensing Module
Product Code. 10749741
Change view
Click to view available options
For Use With (Equipment):
9300BNWP Electrode Body Handle
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10749741 9300BNWP Electrode Body Handle Each
1 options
This item is not returnable. View return policy
Product Code. 10749741 Supplier Thermo Scientific™ Supplier No. 931801
Fecha estimada de envoi: 18-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Get quick, reliable and accurate measurements of dissolved ammonium in aqueous solutions.
The ammonium module must be used with the 9300BNWP electrode body and double junction reference electrode, Cat. No. 900200.
Warranty period
6 months from date of purchase
TRUSTED_SUSTAINABILITY
Specifications
Connector Types WPBNC
For Use With (Equipment) 9300BNWP Electrode Body Handle
Type Ammonia Ion Sensing Module