missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Modular Block Accessories

Accessories for modular block dry baths

Marca:  Thermo Scientific™ 2064Q

Detalles adicionales : Peso : 0.99000kg

 Ver más versiones de este producto

Código de producto. 10626304

  • 369.00€ / cada uno


Consúltenos : 902 239 303 
Añadir a la cesta

Includes: Thermometer well for monitoring block temperatures; block puller for safe, easy block removal

Descripción

Descripción

  • For use with the digital 2000 series and analog 2050 series modular block dry baths
  • Designed to provide close contact of tubes to block walls for excellent heat transfer
  • Block W x H x D: 9.5 x 5.1 x 7.4cm (3.75 x 2 x 2.9 in.)

Especificaciones

Especificaciones

Lab-Line modular dri-bath
Black
Digital 2000 series and analog 2050 series
51mm
Aluminum
74mm
Modular Block
Thermometer and block puller
Videos