Learn More
Thermo Scientific™ Paquetes de aplicación de reómetro HAAKE™ Viscotester™ iQ
Listo para su uso en cuestión de segundos en cualquier configuración que necesite
Marca:  Thermo Scientific™ 262-0005
Detalles adicionales : Peso : 4.90000kg
 Ver más versiones de este producto
Código de producto. 15325123
  • 12400.00€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
Acquire simple viscosity curves or perform complex rheological investigations covering measurements of low viscous liquids up to highly viscous pastes with the Thermo Scientific™ HAAKE™ Viscotester™ rheometer, which can be used as a standalone unit or optionally, be controlled by software. The size and footprint of the HAAKE™ Viscotester™ rheometer make it a portable unit suitable for mobile use or allow it to serve as an important tool in the quality control laboratory.
Application packages are designed to perform a diverse set of rheological tasks and meet a variety of application needs.
  • Advanced design with one-hand lift operation
  • Touchscreen control panel with multilingual, graphical user interface
  • Data quality assessment and QC quality criteria
  • Connect Assist quick coupling and automatic recognition of temperature modules and rotors
  • Liquid temperature module uses a circulator for coaxial cylinders and parallel plates
  • Self-sufficient Peltier temperature module for coaxial cylinders and parallel plates
  • Small dimensioned, coaxial cylinders use less sample and shorter temperature equilibration times to minimize costs per measurement
  • Universal container holder for relative measurements in the original containers
Especificaciones
Especificaciones
Alimentos, pinturas, tintes, revestimientos
Reómetro HAAKE Viscotester iQ, módulo de temperatura de líquido TM-LI-C32, recipiente CCB25 DIN y rotor CCB25 DIN/Ti
Paquete de aplicaciones para líquidos
Videos