Learn More
Thermo Scientific™ Placa de PCR ABgene™ SuperPlate de 96 pocillos
Placas para PCR con semifaldón de 96 pocillos con una plataforma plana para mejorar el sellado en aplicaciones de PCR y qPCR.
Marca:  Thermo Scientific™ AB-2400/R
Detalles adicionales : Peso : 0.71000kg
 Ver más versiones de este producto
Código de producto. 10113853
  • 141.00€ / paquete de 25
Fecha de envío estimada
del proveedor 31-08-2020
Añadir a la cesta
Descripción
Descripción
The Thermo Scientific 96-Well Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers and ABI platforms, including sequencers, with no adapters necessary. The Thermo Scientific PCR plastics are designed, manufactured, and tested to ensure PCR performance. The plates have 0.3 mL maximum well volume when used with adhesive and heat seals.
The flat plate deck design ensures easy handling, and the plates feature a raised rim to prevent evaporation. The 96-Well Semi-Skirted Flat Deck PCR Plates have a cut corner at the A12 location and have thin well walls for consistent heat transfer.
Highlights
• Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
• Flat plate deck facilitates sealing and handling
• Available as SuperPlate providing 4x more rigidity for superior robotic handling
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: A12
Applications
• PCR
• qPCR/Real-Time PCR
• Sequencing
Hazardous:No
>Storage Condition:Ambient
Especificaciones
Especificaciones
No Barcode
Red
Polypropylene
Semi
300 μL
96
DNA, DNase, RNase-free
For improved sealing in PCR and qPCR applications
Yes
Flat Deck
Standard
8 x 12