Learn More
Alfa Aesar™ Tetramethylammonium nitrate, 98%
Marca:  Alfa Aesar™ 041790.09
Detalles adicionales : CAS : 1941-24-8 Peso : 0.01000kg Transport : UN number : 1479 Chem class : 5.1 Pack group : III
Cantidad. 10g
 Ver más versiones de este producto
Código de producto. 11318948
  • 21.40€ / 10 g
Fecha de envío estimada
del proveedor 25-06-2019
Añadir a la cesta
Especificaciones
1941-24-8
136.151
QGPVYHSXZFOSRL-UHFFFAOYSA-N
74743
C[N+](C)(C)C.[N+](=O)([O-])[O-]
C4H12N2O3
MFCD00043103
tetramethylammonium nitrate, methanaminium, n,n,n-trimethyl-, nitrate, tetramethylazanium nitrate, methanaminium, n,n,n-trimethyl-, nitrate 1:1, acmc-20ajj7, tetramethylammonium nitrat, tetra methyl ammonium nitrate, ksc174c1t, tetramethylammonium nitrate, tetramethylammonium nitrate
tetramethylazanium;nitrate
Especificaciones
Tetramethylammonium nitrate
>345°C
1941-24-8
MFCD00043103
tetramethylammonium nitrate, methanaminium, n,n,n-trimethyl-, nitrate, tetramethylazanium nitrate, methanaminium, n,n,n-trimethyl-, nitrate 1:1, acmc-20ajj7, tetramethylammonium nitrat, tetra methyl ammonium nitrate, ksc174c1t, tetramethylammonium nitrate, tetramethylammonium nitrate
C[N+](C)(C)C.[N+](=O)([O-])[O-]
136.151
136.15
98%
Odorless
10g
C4H12N2O3
UN1479
QGPVYHSXZFOSRL-UHFFFAOYSA-N
tetramethylazanium;nitrate
74743
Crystalline