missing translation for 'onlineSavingsMsg'
Learn More

Cloruro de tetraetilamonio monohidrato, +98 %, Thermo Scientific Chemicals

Catalog Number 11377358
Click to view available options
Cantidad:
50 g
250 g
Este artículo no se puede devolver. Vea la política de devoluciones
68696-18-4
C8H22ClNO
183.72
MFCD00149992
CALLTGJPWMIDPC-UHFFFAOYSA-M
tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate
16211205
tetraetilazanio; cloruro; hidrato
CC[N+](CC)(CC)CC.O.[Cl-]
Este artículo no se puede devolver. Vea la política de devoluciones
68696-18-4
C8H22ClNO
183.72
MFCD00149992
CALLTGJPWMIDPC-UHFFFAOYSA-M
tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate
16211205
tetraetilazanio; cloruro; hidrato
CC[N+](CC)(CC)CC.O.[Cl-]

Catalizador de transferencia de fase

Phase transfer catalystTetraethylammonium chloride monohydrate is used in the preparation of 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl chloride by reacting with 1,2-O-acetoxonium-3,4,6-tri-O-acetyl-alfaD-glucopyranose perchlorate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Tetraethylammonium chloride monohydrate
Punto de fusión ∼110°C
Densidad 1.0801 g/mL
Cantidad 250 g
Beilstein 3563247
Índice Merck 14,9200
Información de solubilidad Soluble in water.
Formula Weight (peso de la fórmula) 183.72 (165.71 Anhydrous)
Porcentaje de pureza ≥98%
Forma física cristalino/a/s

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'