Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint without Graduation
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca:  Lenz Laborglasintrumente 05700114
Detalles adicionales : Peso : 0.10000kg
Código de producto. 10018170
Cantidad Precio
1 5.52€ / cada uno
10 4.62€ / cada uno
Envío estimado: 02-03-2022
Añadir a la cesta
Descripción
Descripción
  • With round bottom, with NS socket
  • Available without stopper
  • With PE stopper or with hexagonal hollow glass stopper
  • Made of DURAN™ tubing
Especificaciones
Especificaciones
PE Stopper
16 mm
160 mm