Learn More
OHAUS™ Test Tube Rack
Product Code. 15732529 Shop All Ohaus Products
Change view
Click to view available options
Positions:
12
17
21
24
33
35
40
48
60
63
90
Holds:
Tubes (10 to 13 mm)
Tubes (10 to 14 mm)
Tubes (14 to 16 mm)
Tubes (15 mL)
Tubes (16 mm)
Tubes (16 to 20 mm)
Tubes (18 to 20 mm)
Tubes (20 mm)
Tubes (21 to 25 mm)
Tubes (22 to 25 mm)
Tubes (25 mm)
Tubes (30 mm)
Tubes (5 mL)
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Holds unitSize
15732529 33 Tubes (16 to 20 mm) Each
15722539 12 Tubes (5 mL) Each
15752529 17 Tubes (5 mL) Each
15742529 21 Tubes (21 to 25 mm) Each
15712529 21 Tubes (30 mm) Each
15702539 24 Tubes (22 to 25 mm) Each
15702529 24 Tubes (25 mm) Each
15712539 35 Tubes (15 mL) Each
15792529 35 Tubes (18 to 20 mm) Each
15792519 40 Tubes (20 mm) Each
15722529 48 Tubes (10 to 14 mm) Each
15782529 48 Tubes (14 to 16 mm) Each
15782519 60 Tubes (16 mm) Each
15772529 63 Tubes (10 to 13 mm) Each
15772519 90 Tubes (30 mm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 15732529 Supplier OHAUS™ Supplier No. 30400111
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
OHAUS™ Test Tube Rack designed to securely hold tubes in place.
  • Constructed of stainless steel
  • For use with tubes of various sizes
TRUSTED_SUSTAINABILITY
Specifications
Description Test Tube Rack 16-20 mm Stationary
Autoclavable Autoclavable
Color Blue
Holds Tubes (16 to 20 mm)
Material PVC-coated Steel
Positions 33
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Model Stationary
Size 16 to 22 mm
Dimensions (L x W x H) 9.5 x 42 x 10.2 cm
Length (Metric) 9.50 cm
Length (English) 3.75 in.
Width (English) 16.50 in.
Height (English) 4.00 in.
Depth (English) 3.75 in.
Depth (Metric) 9.50 cm
Diameter (Metric) Opening 20 mm
Product Type Test Tube Rack
Shelves 3 Tiers
Height (Metric) 10.200 cm
Array 3 x 11
Width (Metric) 42.00 cm