Learn More
STEMCELL Technologies TeSR™-E8™3D
Animal component-free suspension culture medium for human ES and iPS cells
Marca: STEMCELL Technologies 03990
Detalles adicionales : Peso : 0.60000kg
Descripción
- TeSR™-E8™3D is an animal component-free culture medium developed for the expansion and scale-up of undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells as aggregates in 3D suspension culture
- Based on TeSR™-E8™, TeSR™-E8™3D is a low-protein medium with an optimized protocol for scaling up human ES and iPS cell production in suspension culture
- The novel fed-batch workflow saves time and media; daily feeds replenish nutrients and eliminate the need for medium exchanges on non-passaging days
- TeSR™-E8™3D is made with the highest level of quality and care
- It contains only the essential components required for scale-up of ES and iPS cells
- The TeSR™-E8™3D system is compatible with cells previously cultured in mTeSR™1 or TeSR™-E8™ on Corning™ Matrigel™ hESC-Qualified Matrix (Corning Catalog #354277) or Vitronectin XF™ (Catalog #07180, a matrix developed and manufactured by Nucleus Biologics), and is compatible with a range of suspension culture vessels
Especificaciones
Pluripotent Stem Cells | |
Cell Culture, Expansion | |
1 Kit |