missing translation for 'onlineSavingsMsg'
Learn More

TES Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10258-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816533

  • 109.00€ / 20 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

TES maps to a common fragile site on chromosome 7q31. 2 designated FRA7G and this gene is similar to mouse Testin, a testosterone-responsive gene encoding a Sertoli cell secretory protein containing three LIM domains. LIM domains are double zinc-finger motifs that mediate protein-protein interactions between transcription factors, cytoskeletal proteins and signaling proteins. Multiple protein isoforms are encoded by transcript variants of the TES gene and Cancer-associated chromosomal changes often involve regions containing fragile sites involving the TES gene.
Especificaciones

Especificaciones

TES
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P47226, Q2LAP6, Q9UGI8
TES, Testin
Rabbit
IgG
20 μL
-20°C
Primary
21753, 26136, 500040
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
TES
Liquid
Tes
TES Fusion Protein Ag0379
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documentos
Certificados