Learn More
terc-Butilo 4-(3-aminobencil)piperazin-1-carboxilato, 97 %, Thermo Scientific™
Catalog Number p-4510392
Click to view available options
Cantidad:
250 mg
1 g
5 g
361345-40-6
C16H25N3O2
291.395
VKSINZNBWVJILC-UHFFFAOYSA-N
tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
7127799
terc-butilo 4-[(3-aminofenil)metil]piperazin-1-carboxilato
CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N
361345-40-6
C16H25N3O2
291.395
VKSINZNBWVJILC-UHFFFAOYSA-N
tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
7127799
terc-butilo 4-[(3-aminofenil)metil]piperazin-1-carboxilato
CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N
Nombre del producto químico o material tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate
Intervalo de porcentaje de ensayo 97%
Cantidad 250 mg
Formula Weight (peso de la fórmula) 291.39
Porcentaje de pureza 97%