Learn More
TER-119 Rat anti-Mouse, Super Bright 702, Clone: TER-119, eBioscience™
Rat Monoclonal Antibody
Marca:  Invitrogen 67-5921-82
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15913774
  • 279.00€ / 100 microgramos
Fecha de envío estimada
del proveedor 01-08-2019
Añadir a la cesta
Descripción
Descripción
When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 702 nm; Laser: Violet Laser TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.
Especificaciones
Especificaciones
TER-119
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
TER-119
Super Bright 702
Ly76
Ly76
IgG2b, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
104231