missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TELO2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15975-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TELO2 is a protein that functions as an S-phase checkpoint protein in the cell cycle. The protein may also play a role in DNA repair.Especificaciones
TELO2 | |
Polyclonal | |
Unconjugated | |
TELO2 | |
CLK2, hCLK2, KIAA0683, Protein clk 2 homolog, TEL2, TELO2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9894 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y4R8 | |
TELO2 | |
TELO2 Fusion Protein Ag8763 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |