Learn More
Ansell Edmont™ Marigold™ Foodsure Tear-Resistant Natural Rubber Gloves
Click to view available options
Tamaño:
7.5
Large
Medium
Small
Descripción
- Industrial protective glove
- Strong yet sensitive formulation
- Unsupported flocked natural rubber glove
- 100% cotton flock for good comfort
- Negative pattern grip finish on palm and fingers for good dry grip
- Good tear resistance
- Tear resistant beaded cuff for easy donning
- Not recommended for fatty food
Applications:
- Food preparation, processing and handling
- Meat and poultry processing
- Fish processing
Especificaciones
Certificaciones/conformidad | 2010 , AQL 1.5 |
Color | Blue |
Latex Free | No |
Material | Latex |
Tamaño | 7.5 |
Grosor (métrico) | 0.43 mm |