Learn More
TdT Mouse anti-Human, FITC, Clone: TdT-6, Invitrogen™
Mouse Monoclonal Antibody
Marca:  Invitrogen MHTDT012
Detalles adicionales : Peso : 0.50000kg
 Ver más versiones de este producto
Código de producto. 10785723
  • 319.00€ / cada uno
Stock EU 2
Añadir a la cesta
Descripción
Descripción
Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3′ hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt′s lymphoma.
Especificaciones
Especificaciones
TdT
Monoclonal
FITC
DNTT
Liquid
DNTT
Human Terminal Deoxynucleotidyl Transferase (TdT)
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
TdT-6
PBS with 1% BSA and 0.1% sodium azide
P04053
TDT, DNA nucleotidylexotransferase, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, terminal addition enzyme, terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, terminal transferase
Mouse
IgG1
0.2 mL
4° C, store in dark
Primary
1791