missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25239-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TDRD5, also named as Tudor domain-containing protein 5, is a 981 amino acid protein, which contains one Tudor domain and three HTH OST-type domains. TDRD5 localizes in the cytoplasm and belongs to the TDRD5 family. TDRD5 is required during spermiogenesis to participate in the repression transposable elements and prevent their mobilization, which is essential for the germline integrity.Especificaciones
TDRD5 | |
Polyclonal | |
Unconjugated | |
TDRD5 | |
RP11 427G13.1, TDRD5, tudor domain containing 5, TUDOR3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
163589, 214575 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VCS6, Q8NAT2 | |
Tdrd5 | |
TDRD5 Fusion Protein Ag18896 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |