missing translation for 'onlineSavingsMsg'
Learn More

TCEB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10779-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817393

  • 109.00€ / 20 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).
Especificaciones

Especificaciones

TCEB2
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P62869, P62870, Q15370
ELOB, Elongin 18 kDa subunit, Elongin B, SIII, SIII p18, TCEB2
Rabbit
IgG
20 μL
-20°C
Primary
67673, 6923, 81807
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
ELOB
Liquid
Elob
TCEB2 Fusion Protein Ag1228
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documentos
Certificados