missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17825-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TCEA1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated; transcription, DNA-templated.Especificaciones
TCEA1 | |
Polyclonal | |
Unconjugated | |
TCEA1 | |
GTF2S, SII, SII TFIIS, TCEA, TCEA1, TF2S, TFIIS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21399, 362479, 6917 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10711, P23193, Q4KLL0 | |
Tcea1 | |
TCEA1 Fusion Protein Ag12199 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |