missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBL2 Mouse anti-Human, Clone: 1D5C2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66307-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the beta-transducin protein family. Most proteins of the beta-transducin family are involved in regulatory functions. This protein is possibly involved in some intracellular signaling pathway. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.Especificaciones
TBL2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y4P3 | |
TBL2 | |
TBL2 Fusion Protein Ag24225 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgA |
Immunohistochemistry (Paraffin), Western Blot | |
1D5C2 | |
Unconjugated | |
TBL2 | |
TBL2, transducin (beta) like 2, Transducin beta like protein 2, WBSCR13, WS betaTRP | |
Mouse | |
Affinity Chromatography | |
RUO | |
26608 | |
-20°C | |
Liquid |