missing translation for 'onlineSavingsMsg'
Learn More

TBL1XR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 55312-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816805

  • 109.00€ / 20 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation.
Especificaciones

Especificaciones

TBL1XR1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BHJ5, Q9BZK7
C21, DC42, IRA1, TBL1 related protein 1, TBL1XR1, TBLR1
Rabbit
IgG
20 μL
-20°C
Primary
79718, 81004
Western Blot
Unconjugated
TBL1XR1
Liquid
TBL1XR1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
SDS
Documentos
Certificados