missing translation for 'onlineSavingsMsg'
Learn More

TAPT1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 27657-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16815765

  • 319.00€ / 150 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a highly conserved, putative transmembrane protein. A mutation in the mouse ortholog of this gene results in homeotic, posterior-to-anterior transformations of the axial skeleton which are similar to the phenotype of mouse homeobox C8 gene mutants. This gene is proposed to function downstream of homeobox C8 to transduce extracellular patterning information during axial skeleton development. An alternatively spliced transcript variant encoding a substantially different isoform has been described, but its biological validity has not been determined.
Especificaciones

Especificaciones

TAPT1
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6NXT6
CMVFR, TAPT1
Rabbit
IgG
150 μL
-20°C
Primary
202018
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TAPT1
Liquid
TAPT1
TAPT1 Fusion Protein Ag26360
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados