Learn More
Tamsulosin hydrochloride, 98+%
Product Code. 15484469
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484469 100 mg 100mg
15464469 10 mg 10mg
15474469 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15484469 Supplier Thermo Scientific Alfa Aesar Supplier No. J61999.MC
 Restricted Item
Fecha estimada de envoi: 15-12-2026
Add to basket
This item is not returnable. View return policy
An alpha-1 adrenoceptor antagonist | CAS: 106463-17-6 | C20H29ClN2O5S | 444.971 g/mol
An alpha-1 adrenoceptor antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106463-17-6
Molecular Formula C20H29ClN2O5S
Molecular Weight (g/mol) 444.971
MDL Number MFCD00922997
InChI Key ZZIZZTHXZRDOFM-XFULWGLBSA-N
Synonym tamsulosin hydrochloride, omnic, tamsulosin hcl, pradif, flomax, urolosin, secotex, josir, alna, omic
PubChem CID 5362376
ChEBI CHEBI:9399
IUPAC Name 5-[(2R)-2-[2-(2-ethoxyphenoxy)ethylamino]propyl]-2-methoxybenzenesulfonamide;hydrochloride
SMILES CCOC1=CC=CC=C1OCCNC(C)CC2=CC(=C(C=C2)OC)S(=O)(=O)N.Cl
Specifications
Melting Point 228°C to 230°C
Color White
Quantity 100 mg
Merck Index 14,9049
Formula Weight 444.97
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tamsulosin hydrochloride