Learn More
Gilac™ Tampico Oven Broom
Product Code. 17988631
Change view
Click to view available options
Width (Metric):
38 cm
50 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
17988631 50 cm Each
17928461 38 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17988631 Supplier Gilac™ Supplier No. G020030
  • ONLINE EXCLUSIVE PRICE
    76.77€
    50.78€ / paquete de 1
    Ahorro 25.99€
    33% Descuento
Fecha estimada de envoi: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Tampico Oven Broom is a special brush for bakery and pastry.
  • Special brush for bakery and pastry.
Specifications
Product Type Broom Head
Color Code Orange
Sterility Non-sterile
Autoclavable Not Autoclavable
Width (Metric) 50 cm
Width (English) 19.7 in.
Color-coded Yes
Material Wood