missing translation for 'onlineSavingsMsg'
Learn More

Talin1 Antibody (TA205), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-21642AF647

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213530

  • 566.94€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Talin1 Monoclonal specifically detects Talin1 in Human, Chicken, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Especificaciones

Especificaciones

Talin1
Monoclonal
Alexa Fluor 647
Q9Y490
TLN1
Human platelet talin.
0.1 mL
Cytoskeleton Markers
7094
Human, Chicken, Rabbit
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
TA205
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
ILWEQ, KIAA1027, talin 1, talin-1, TLN
Mouse
Protein G purified
RUO
Primary
Immunoblots the 225 kDa talin band on SDS gels of platelets and other cells. It may be used to stain adhesion plaques in fixed and permeabilized fibroblasts. TA205 recognizes an epitope within residues 139-433.
Store at 4C in the dark.
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only