missing translation for 'onlineSavingsMsg'
Learn More

TAB2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-81735B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213997

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

TAB2 Polyclonal antibody specifically detects TAB2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones

Especificaciones

TAB2
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
FLJ21885, KIAA0733CHTD2, MAP3K7IP2mitogen-activated protein kinase kinase kinase 7 interacting protein 2, Mitogen-activated protein kinase kinase kinase 7-interacting protein 2, TAB-2, TAK1-binding protein 2, TGF-beta activated kinase 1/MAP3K7 binding protein 2, TGF-beta-activated kinase 1 and MAP3K7-binding protein 2, TGF-beta-activated kinase 1-binding protein 2
TAB2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human TAB2. The immunogen is located within the last 50 amino acids of TAB2. Amino Acid Squence: KTPKTQDTEDDEGA
0.1 mL
Cytokine Research, Signal Transduction
23118
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales