Learn More
CELLPATH SYSTEM II+ SLOTTED CASSETTE
Product Code. 30226242
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 2000
Pack of 3000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226242 Threaded Pack of 2000
30226240 Loose Pack of 1000
30226241 Hoppered Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30226242 Supplier CELLPATH Supplier No. EAM050272B
  • ONLINE EXCLUSIVE PRICE
    334.81€
    251.05€ / paquete de 2000
    Ahorro 83.76€
    25% Descuento
View all promotions
 Restricted Item
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Incorporating 1.1 mm wide slots, the System II+ Slotted Cassette increases reagent flow, facilitating improved consistency and quality of processed specimens. CellPath's System II+ Cassettes come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Threaded
Product Type Cassette