missing translation for 'onlineSavingsMsg'
Learn More

Synuclein alpha/beta Antibody (3B6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-22957

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213240

  • 485.95€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Synuclein alpha/beta Monoclonal specifically detects Synuclein alpha/beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

Synuclein alpha/beta
Monoclonal
1.0 mg/mL
Western Blot 1:1000-1:2000, Flow Cytometry 2-5ug ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100
beta-synuclein, I(2)-synuclein, I+/--synuclein, NACP, non A-beta component of AD amyloid, PARK1, PD1, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein
Mouse
Protein G purified
RUO
Primary
Anti-human alpha-synuclein (alpha-synuclein 119-140aa) mAb, clone 3B6, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human alpha-synuclein protein.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
3B6
Unconjugated
NP_000336
SNCA
This Synuclein alpha/beta Antibody (3B6) was developed against recombinant human Synuclein alpha/beta (119-140aa) purified from E. coli.
0.1 mL
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
6622
Human, Mouse, Rat
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only