Learn More
Pacific Handy Cutter Swivel Holster for RZ-3/EZ-3
Product Code. 17729047
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 10
Product Code. Color Unit Size
17729047 Black Pack of 10
1 options
This item is not returnable. View return policy
Product Code. 17729047 Supplier Pacific Handy Cutter Supplier No. UKH430
  • ONLINE EXCLUSIVE PRICE
    95.80€
    82.97€ / paquete de 10
    Ahorro 12.83€
    13% Descuento
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Practical belt holster made of plastic with fastening clip for belt or clothing. The knife holder can be swivelled.
  • As a result, a comfortable wearing position can be achieved at any time, whether standing or sitting.
  • Suitable for the safety knives EZ-3 and RZ-3.
  • Particularly easy to clean.
Specifications
Product Type Swivel Holster
For Use With (Equipment) Safety Knives EZ-3 and RZ-3
Material Plastic
Color Black
Sterility Non-sterile