Learn More
SVL™ Bearing Stirrer for SVL™ Stirrer Shaft
Designed to accept SVL™ stirrer shafts with ground glass surfaces.
Marca:  SVL 588-08
Detalles adicionales : Peso : 0.07000kg
 Ver más versiones de este producto
Código de producto. 15547730
  • 915.27€ / paquete de 5
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Borosilicate glass complete with fittings
  • Glands manufactured from PTFE based polymer which is self lubricating and corrosion-resistant
  • Two types available: With SVL™ screwthread to allow connection to SVL™ flasks, With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks
Especificaciones
Especificaciones
SVL™ stirrer shaft
Quickfit™ ground Glass cone to allow connection to Quickfit™ flask
PTFE Based Polymer
24/29