missing translation for 'onlineSavingsMsg'
Learn More

SV40 T-antigen Antibody (PAB416), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-59676AF532

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213554

  • 551.82€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

SV40 T-antigen Monoclonal specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Especificaciones

Especificaciones

SV40 T-antigen
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
SV40 transformed mouse cells
Cancer, Virology Bacteria and Parasites
Specific for N-terminal domain of SV40 large and small T-antigen
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
PAB416
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Mouse
0.1 mL
Primary
Virus
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales