Learn More
Hettich Lab Technology™ Suspension Carrier
Product Code. 10740223
Change view
Click to view available options
For Use With (Equipment):
Suspension Swing-Out Rotor 1460, Plates: MTP, CP, DWP, MS, QP, Microtest (Terasaki), PCR plate, PCR strips with plate
Suspension Swing-Out Rotor 1624 Adapter 5277, 1357, 1327, 1326
Suspension Swing-Out Rotor 1624, 4758, 1726 Adapter 5277, 1357, 1327, 1326
Suspension Swing-Out Rotor 1798, 4753, 5694, 4446, 4296
Suspension, Swing-Out Rotor 1624, 1726, 4758
Suspension, Swing-out Rotor 1494
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10740223 Suspension, Swing-out Rotor 1494 Each
10424635 Suspension, Swing-Out Rotor 1624, 1726, 4758 Each
10575705 Suspension Swing-Out Rotor 1624, 4758, 1726 Adapter 5277, 1357, 1327, 1326 Each
10117010 Suspension Swing-Out Rotor 1798, 4753, 5694, 4446, 4296 Each
10049430 Suspension Swing-Out Rotor 1624 Adapter 5277, 1357, 1327, 1326 Each
11942248 Suspension Swing-Out Rotor 1460, Plates: MTP, CP, DWP, MS, QP, Microtest (Terasaki), PCR plate, PCR strips with plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10740223 Supplier Hettich Lab Technology™ Supplier No. 1425
Fecha estimada de envoi: 20-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Suspension Carrier fits a swing-out rotor and may accommodate a variety of buckets and/or tubes.
  • Available in a variety of configurations
  • Configurations capable of accommodating a host of customized centrifuge workloads
Specifications
For Use With (Equipment) Suspension, Swing-out Rotor 1494
Product Type Suspension Carrier
Container Type Swing-out Suspension Carrier
No. of Places Adapter Dependent