Learn More
Survivin Antibody (8E2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 14 publications
Brand:  Novus Biologicals NB500-644SS
Additional Details : Peso : 0.00970kg
 View more versions of this product
Product Code. 18214430
  • 207.34€ / 25 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock
Description
Description
Survivin Monoclonal specifically detects Survivin in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In vitro assay, Flow (Intracellular), Block/Neutralize.
Specifications
Specifications
Survivin
Monoclonal
1 mg/mL
Flow Cytometry 1 ug/mL. Use reported in scientific literature (PMID 11313271), ELISA reported in scientific literature (PMID 9581817), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:100, In vitro assay, Flow (Intracellular), Block/Neutralize reported in scientific literature (PMID 14614132)
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Mouse
16 kDa
0.025 mL
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
332
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
800
Unconjugated
O15392, Q9JHY7
BIRC5
This Survivin Antibody (8E2) was developed against full length recombinant human Survivin [UniProt# O15392]
Protein G purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
For Research Use Only