Learn More
Terumo™ SurGuard2™ Hypodermic Safety Needles
Designed with safety in mind, these hypodermic needles include a simple locking mechanism to reduce needle stick injuries. Terumo™ SurGuard2™ Hypodermic Safety Needles can be easily activated using only one hand.
Marca:  Terumo™ 8SG22516MP
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12661146
  • 61.40€ / paquete de 100
Stock Remoto 11
Añadir a la cesta
Descripción
Descripción
  • Simple locking mechanism for safety
  • Safety lock can be activated using only one hand
  • Standard hub equally fits to all luer lock and luer slip syringes
Especificaciones
Especificaciones
25 G
16mm
Hypodermic Needle
Safety lock
800/Pk.