missing translation for 'onlineSavingsMsg'
Learn More

SURF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 16522-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816184

  • 109.00€ / 20 microlitros

Envío estimado: 28-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene shares a bidirectional promoter with surfeit 1 (SURF1; GeneID: 6834), which is located on the opposite strand. It encodes a conserved protein that is expressed in a variety of tissues.
Especificaciones

Especificaciones

SURF2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09926, Q15527
SURF 2, SURF2, surfeit 2, Surfeit locus protein 2
Rabbit
IgG
20 μL
-20°C
Primary
20931, 619345, 6835
Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
SURF2
Liquid
Surf2
SURF2 Fusion Protein Ag9745
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados