Learn More
Hellma™ Suprasil Semi Micro Flow Through Cell
Product Code. 10743091
Change view
Click to view available options
Wavelength Range:
200 to 2500 nm
Unit Size:
Each
Product Code. Wavelength Range Unit Size
10743091 200 to 2500 nm Each
1 options
This item is not returnable. View return policy
Product Code. 10743091 Supplier Hellma™ Supplier No. 176700151040
  • ONLINE EXCLUSIVE PRICE
    555.00€
    504.00€ / paquete de 1
    Ahorro 51.00€
    9% Descuento
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type Semi-micro Flow-through Cell
Capacity (Metric) 390 μL
Material Suprasil
Wavelength Range 200 to 2500 nm
Height (Metric) 35 mm