missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11376-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
SULT1A4 | |
Polyclonal | |
Unconjugated | |
SULT1A4 | |
Aryl sulfotransferase 1A3/1A4, HAST3, M PST, ST1A3/ST1A4, STM, Sulfotransferase 1A3/1A4, SULT1A3, SULT1A4, TL PST | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
445329 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0DMN0 | |
SULT1A4 | |
SULT1A4 Fusion Protein Ag1935 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |