missing translation for 'onlineSavingsMsg'
Learn More

Ácido sulfúrico, solución estandarizada de 0,02 N, Thermo Scientific Chemicals

Catalog Number 11313797
Click to view available options
Cantidad:
1 L
4 L
20 L
Este artículo no se puede devolver. Vea la política de devoluciones
7664-93-9, 7732-18-5
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
ácido sulfúrico
OS(O)(=O)=O
Este artículo no se puede devolver. Vea la política de devoluciones
7664-93-9, 7732-18-5
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
ácido sulfúrico
OS(O)(=O)=O

Se utiliza para análisis volumétricos en laboratorios.

Sulfuric acid, 0.02N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Sulfuric acid
Cantidad 4 L
Formula Weight (peso de la fórmula) 98.08
Solubilidad Miscible with water
Forma física Líquido

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'