Learn More
Sulfur Standard: S @ 500 μg/g (0.0500 wt%) in 75 cSt Mineral Oil, VHG™
Product Code. 16449669
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449669 100 mL 100mL
17910436 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16449669 Supplier VHG Labs Supplier No. VHGSMIN500100
 Restricted Item
Fecha estimada de envoi: 11-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfur Standard: S @ 500 μg/g (0.0500 wt%) in 75 cSt Mineral Oil
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfur
CAS 7704-34-9
Methods XRF
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/S
SMILES [S]
IUPAC Name sulfur
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 32.065
Quantity 100 mL
Formula Weight 31.972 g/mol