Learn More
Sulfamethoxazole-d4, TRC
Product Code. 30427240
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30427240 25 mg 25mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30427240 Supplier Toronto Research Chemicals Supplier No. TRCS69908725MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfamethoxazole-d4
CAS 1020719-86-1
Purity Grade Notes HPLC
Molecular Formula C10 2H4 H7 N3 O3 S
Quantity 25 mg
Synonym Sulfamethoxazole D4 (benzene D4), Benzene-2,3,5,6-d4-sulfonamide','4-amino-N-(5-methyl-3-isoxazolyl)-, 4-Amino-N-(5-methyl-3-isoxazolyl)benzene-2,3,5,6-d4-sulfonamide
InChI Formula InChI=1S/C10H11N3O3S/c1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h2-6H,11H2,1H3,(H,12,13)/i2D,3D,4D,5D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2cc(C)on2
IUPAC Name 4-amino-2,3,5,6-tetradeuterio-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
Molecular Weight (g/mol) 257.3
Formula Weight 257.0772
Percent Purity >95