missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12260-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Heparan sulfate 6-O-endosulfatases, such as Sulfatase 2, selectively remove 6-O-sulfate groups from heparan sulfate. Heparan sulfate proteoglycans (HSPGs) act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules (Dai et al., 2005 ).Especificaciones
SULF2 | |
Polyclonal | |
Unconjugated | |
SULF2 | |
Extracellular sulfatase Sulf 2, HSULF 2, KIAA1247, SULF2, sulfatase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
311642, 55959, 72043 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CFG0, Q8IWU5 | |
Sulf2 | |
SULF2 Fusion Protein Ag2915 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |