Learn More
Novus Biologicals™ SUG1 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-14892
Detalles adicionales : Peso : 0.00970kg
Product Code. 18214524
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock
Description
Description
The SUG1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC21251 and based on accession number NM_285. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens proteasome (prosome, macropain) 26S subunit, ATPase, 5 (PSMC5), mRNA.
Native
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only